Dicyclohexyl ketone ≥97.0%

Leverantör: TCI EUROPE

Synonyms: Dodecahydrobenzophenone

D2258-25ML D2258-500ML
TCIAD2258-25MLEA 230 SEK
TCIAD2258-25ML TCIAD2258-500ML
Dicyclohexyl ketone ≥97.0%
Dicyclohexyl ketone
Formel: C₁₃H₂₂O
MW: 194,32 g/mol
Kokpunkt: 135°C
MDL Number: MFCD00040417
CAS nummer: 119-60-8
EINECS: 204-336-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c14869c077b65d2690f69967f7678950c470462&vid=4c14869c077b65d2690f69967f7678950c470462&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR