Isopropyl propionate ≥99.0%
Leverantör: TCI
|
Fara
|
P0507-25ML
P0507-500ML
TCIAP0507-25MLEA
325
SEK
TCIAP0507-25ML
TCIAP0507-500ML
Isopropyl propionate ≥99.0%
Isopropyl propionate
Formel:
C₆H₁₂O₂ Kokpunkt: 108°C Flampunkt: 15°C |
MDL Number:
MFCD00051553 CAS nummer: 637-78-5 EINECS: 211-300-8 UN: 2409 ADR: 3,II |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...