1-Chloroethylethyl carbonate ≥98.0%

Leverantör: TCI
Varning

C1056-25G C1056-100G
TCIAC1056-25GEA 445 SEK
TCIAC1056-25G TCIAC1056-100G
1-Chloroethylethyl carbonate ≥98.0%
1-Chloroethylethyl carbonate
Formel: C₅H₉ClO₃
Kokpunkt: 160°C
Flampunkt: 62°C
MDL Number: MFCD00040523
CAS nummer: 50893-36-2
EINECS: 256-832-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f794a38142f70922dc28a8c773bdd126d17b015d&vid=f794a38142f70922dc28a8c773bdd126d17b015d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR