1-Adamantanol ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
1-Tricyclo[3.3.1.1(3,7)]decanol, 1-Hydroxytricyclo[3.3.1.1(3,7)]decane, Tricyclo[3.3.1.1(3,7)]decan-1-ol, Adamantan-1-ol
A0939-25G
A0939-250G
TCIAA0939-25GEA
590
SEK
TCIAA0939-25G
TCIAA0939-250G
1-Adamantanol ≥98.0%
1-Adamantanol
Formel:
C₁₀H₁₆O MW: 152,24 g/mol Kokpunkt: 287 °C (1013 hPa) Smältpunkt: 280°C Densitet: 1,06 g/cm³ (25 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00074729 CAS nummer: 768-95-6 EINECS: 212-202-8 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...