3-Methylthiophene ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
M0440-25G
M0440-500G
TCIAM0440-25GEA
470
SEK
TCIAM0440-25G
TCIAM0440-500G
3-Methylthiophene ≥98.0%
3-Methylthiophene
Formel:
C₅H₆S MW: 98,17 g/mol Kokpunkt: 115°C Smältpunkt: –69 °C Densitet: 1,02 g/cm³ (20 °C) Flampunkt: 11°C Storage Temperature: Omgivande |
MDL Number:
MFCD00005470 CAS nummer: 616-44-4 EINECS: 210-482-6 UN: 1993 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...