3-Iodothiophene ≥95.0%

Leverantör: TCI EUROPE
Varning

I0380-1G I0380-5G
TCIAI0380-1GEA 325 SEK
TCIAI0380-1G TCIAI0380-5G
3-Iodothiophene ≥95.0%
3-Iodothiophene
Formel: C₄H₃IS
MW: 210,04 g/mol
Kokpunkt: 75°C
Flampunkt: 78°C
Storage Temperature: Kylskåp
MDL Number: MFCD00037901
CAS nummer: 10486-61-0
EINECS: 234-009-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9555b2248ad5aea455d3427148c4d32e4c627b13&vid=9555b2248ad5aea455d3427148c4d32e4c627b13&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR