2,3,5-Tribromothiophene ≥98.0%
Leverantör: TCI EUROPE
T0956-25G
T0956-250G
TCIAT0956-25GEA
670
SEK
TCIAT0956-25G
TCIAT0956-250G
2,3,5-Tribromothiophene ≥98.0%
2,3,5-Tribromothiophene
Formel:
C₄HBr₃S MW: 320,83 g/mol Kokpunkt: 80 °C (0,5 -rr) Smältpunkt: 29 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00014521 CAS nummer: 3141-24-0 EINECS: 221-544-7 ADR: 6.1,III |
Specifikation testresultat
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...