2,5-Lutidine ≥98.0%
Leverantör: TCI
|
Varning
|
L0065-25ML
L0065-5ML
L0065-500ML
TCIAL0065-25MLEA
870
SEK
TCIAL0065-25ML
TCIAL0065-5ML
TCIAL0065-500ML
2,5-Lutidine ≥98.0%
2,5-Lutidine
Formel:
C₇H₉N Kokpunkt: 157°C Smältpunkt: -6°C Densitet: 0,9346 g/cm³ (15 °C) Flampunkt: 48°C |
MDL Number:
MFCD00006343 CAS nummer: 589-93-5 EINECS: 209-666-9 UN: 1993 ADR: 3,III |
|
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...