2,4-Lutidin ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
2,4-Dimethylpyridine
L0085-25ML
L0085-500ML
TCIAL0085-25MLEA
215
SEK
TCIAL0085-25ML
TCIAL0085-500ML
2,4-Lutidin ≥98.0%
2,4-Lutidin
Formel:
C₇H₉N MW: 107,16 g/mol Kokpunkt: 157°C Smältpunkt: -64°C Densitet: 0,93 g/cm³ (20 °C) Flampunkt: 37°C Storage Temperature: Omgivande |
MDL Number:
MFCD00006337 CAS nummer: 108-47-4 EINECS: 203-586-8 UN: 1992 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...