4-(3-Phenylpropyl)pyridine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
P0218-25G
P0218-500G
TCIAP0218-25GEA
215
SEK
TCIAP0218-25G
TCIAP0218-500G
4-(3-Phenylpropyl)pyridine ≥98.0%
4-(3-Phenylpropyl)pyridine
Formel:
C₁₄H₁₅N MW: 197,28 g/mol Kokpunkt: 322 °C (1013 hPa) Densitet: 1,0224 g/cm³ (20 °C) Flampunkt: 136°C |
MDL Number:
MFCD00047458 CAS nummer: 2057-49-0 EINECS: 218-159-1 |
Specifikation testresultat
Appearance | Colorless to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...