3-Etylpyridin ≥98.0%
Leverantör: TCI
|
Varning
|
E0169-25ML
E0169-5ML
TCIAE0169-25MLEA
1725
SEK
TCIAE0169-25ML
TCIAE0169-5ML
3-Etylpyridin ≥98.0%
3-Etylpyridin
Formel:
C₇H₉N Kokpunkt: 165°C Flampunkt: 48°C |
MDL Number:
MFCD00006413 CAS nummer: 536-78-7 EINECS: 208-647-2 UN: 1993 ADR: 3,III Merck Index: 14,03848 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...