2-Etylpyridin ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

E0168-25ML E0168-500ML E0168-100ML
TCIAE0168-25MLEA 470 SEK
TCIAE0168-25ML TCIAE0168-500ML TCIAE0168-100ML
2-Etylpyridin ≥98.0% (by GC, titration analysis)
2-Etylpyridin
Formel: C₇H₉N
Kokpunkt: 144°C
Smältpunkt: –63 °C
Densitet: 0,937 g/cm³ (20 °C)
Flampunkt: 29°C
Storage Temperature: Omgivande
MDL Number: MFCD00006361
CAS nummer: 100-71-0
EINECS: 202-881-9
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c306401a560ca62362ddd973c8c1bd6c47b5f10&vid=2c306401a560ca62362ddd973c8c1bd6c47b5f10&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR