2-Bromo-6-methylpyridine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
6-Bromo-2-picoline, 2-Bromo-6-picoline
B2114-25G
B2114-5G
TCIAB2114-25GEA
1550
SEK
TCIAB2114-25G
TCIAB2114-5G
2-Bromo-6-methylpyridine ≥98.0% (by GC, titration analysis)
2-Bromo-6-methylpyridine
Formel:
C₆H₆BrN MW: 172,02 g/mol Kokpunkt: 102°C Flampunkt: 97°C |
MDL Number:
MFCD00040743 CAS nummer: 5315-25-3 EINECS: 226-173-4 |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...