2-Bromo-5-methylpyridine ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Bromo-5-picoline

B2113-25G B2113-5G
TCIAB2113-25GEA 1975 SEK
TCIAB2113-25G TCIAB2113-5G
2-Bromo-5-methylpyridine ≥98.0% (by HPLC, titration analysis)
2-Bromo-5-methylpyridine
Formel: C₆H₆BrN
MW: 172,02 g/mol
Kokpunkt: 65 °C (0,5 -rr)
Smältpunkt: 152,5 °C
MDL Number: MFCD00209553
CAS nummer: 3510-66-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9a085c619c1e21f86c8c22d6c428e75a0e7e817&vid=e9a085c619c1e21f86c8c22d6c428e75a0e7e817&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 41.0 to 46.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR