3-Ethoxy-2-nitropyridine ≥98.0%
Leverantör: TCI
|
Varning
|
E0390-1G
E0390-5G
TCIAE0390-1GEA
495
SEK
TCIAE0390-1G
TCIAE0390-5G
3-Ethoxy-2-nitropyridine ≥98.0%
3-Ethoxy-2-nitropyridine
Formel:
C₇H₈N₂O₃ Kokpunkt: 175°C Smältpunkt: 31…32 °C |
MDL Number:
MFCD00006257 CAS nummer: 74037-50-6 EINECS: 277-676-0 ADR: 4.1,II |
|
Specifikation testresultat
Purity (GC) | min. 98.0 % |
Freezing point | 25.0 to 28.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...