2,3-Diaminopyridine ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: Pyridine-2,3-diyldiamine
D0117-25G D0117-5G
TCIAD0117-25GEA 5825 SEK
TCIAD0117-25G TCIAD0117-5G
2,3-Diaminopyridine ≥98.0%
2,3-Diaminopyridine
Formel: C₅H₇N₃
MW: 109,13 g/mol
Kokpunkt: 148 - 150 °C (5 -rr)
Smältpunkt: 110…112 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006319
CAS nummer: 452-58-4
EINECS: 207-200-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94ec6a0fb64bbf11f30d483205b8a5d24316ceb9&vid=94ec6a0fb64bbf11f30d483205b8a5d24316ceb9&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 112.0 to 119.0 deg-C
Solubility in Water almost transparency