4-Aminopyridine ≥99.0% (by GC, titration analysis)
Leverantör: TCI
Fara
A0414-25G A0414-500G A0414-100G
TCIAA0414-25GEA 325 SEK
TCIAA0414-25G TCIAA0414-500G TCIAA0414-100G
4-Aminopyridine ≥99.0% (by GC, titration analysis)
4-Aminopyridine
Formel: C₅H₆N₂
Kokpunkt: 287°C
Smältpunkt: 157…160 °C
Densitet: 1,25 g/cm³ (20 °C)
Flampunkt: 156 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006439
CAS nummer: 504-24-5
EINECS: 207-987-9
UN: 2671
ADR: 6.1,II
Merck Index: 14,03933
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9af5cbcacf6af70e3eb96565ed53ef0cd81fd83&vid=b9af5cbcacf6af70e3eb96565ed53ef0cd81fd83&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 157.0 to 162.0 deg-C