3-Aminopyridine ≥99.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Pyridin-3-amine, 3-Pyridylamine
A0413-25G
A0413-500G
A0413-100G
TCIAA0413-25GEA
285
SEK
TCIAA0413-25G
TCIAA0413-500G
TCIAA0413-100G
3-Aminopyridine ≥99.0% (by GC, titration analysis)
3-Aminopyridine
Formel:
C₅H₆N₂ MW: 94,12 g/mol Kokpunkt: 248°C Smältpunkt: 60…63 °C Flampunkt: 88 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00006400 CAS nummer: 462-08-8 EINECS: 207-322-2 UN: 2671 ADR: 6.1,II Merck Index: 14,00473 |
Specifikation testresultat
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 99.0 % |
Melting point | 59.0 to 65.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...