3-Fluoropyridine ≥98.0%
Leverantör: TCI
|
Fara
|
F0207-25G
F0207-5G
TCIAF0207-25GEA
1225
SEK
TCIAF0207-25G
TCIAF0207-5G
3-Fluoropyridine ≥98.0%
3-Fluoropyridine
Formel:
C₅H₄FN Kokpunkt: 106°C Densitet: 1,13 g/cm³ (20 °C) Flampunkt: 13°C Storage Temperature: Omgivande |
MDL Number:
MFCD00006374 CAS nummer: 372-47-4 EINECS: 206-755-4 UN: 1993 ADR: 3,II |
|
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...