2-Chloro-3,5-dinitropyridine ≥98.0%

Leverantör: TCI EUROPE
Fara

C0943-1G C0943-5G
TCIAC0943-1GEA 525 SEK
TCIAC0943-1G TCIAC0943-5G
2-Chloro-3,5-dinitropyridine ≥98.0%
2-Chloro-3,5-dinitropyridine
Formel: C₅H₂ClN₃O₄
MW: 203,54 g/mol
Smältpunkt: 62…65 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006233
CAS nummer: 2578-45-2
EINECS: 219-937-3
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=740c88737ebff307da9d7e0a8f1a12980f51e5d2&vid=740c88737ebff307da9d7e0a8f1a12980f51e5d2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 66.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR