2,4-Dichloropyridine ≥98.0%
Leverantör: TCI
|
Varning
|
D3784-25G
D3784-5G
TCIAD3784-25GEA
2450
SEK
TCIAD3784-25G
TCIAD3784-5G
2,4-Dichloropyridine ≥98.0%
2,4-Dichloropyridine
Formel:
C₅H₃Cl₂N Kokpunkt: 190°C Flampunkt: 89°C |
MDL Number:
MFCD00955618 CAS nummer: 26452-80-2 EINECS: 247-717-7 ADR: 6.1,III |
|
Specifikation testresultat
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...