2,3,5,6-Tetrachloropyridine 98,0 GC_ANALYSMETOD
Leverantör: TCI
|
Varning
|
T2331-25G
T2331-5G
TCIAT2331-25GEA
535
SEK
TCIAT2331-25G
TCIAT2331-5G
2,3,5,6-Tetrachloropyridine 98,0 GC_ANALYSMETOD
2,3,5,6-Tetrachloropyridine
Formel:
C₅HCl₄N MW: 216.88 g/mol Kokpunkt: 251°C |
MDL Number:
MFCD00092428 CAS nummer: 2402-79-1 EINECS: 219-283-9 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 91.0 to 95.0 deg-C |