2-Bromo-5-nitropyridine ≥98.0%

Leverantör: TCI
Varning

B2321-1G B2321-5G
TCIAB2321-1GEA 215 SEK
TCIAB2321-1G TCIAB2321-5G
2-Bromo-5-nitropyridine ≥98.0%
2-Bromo-5-nitropyridine
Formel: C₅H₃BrN₂O₂
Kokpunkt: 147°C
MDL Number: MFCD00006222
CAS nummer: 4487-59-6
EINECS: 224-777-2
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db3679f98da6c61a703718f4bf152b4b6d326fe4&vid=db3679f98da6c61a703718f4bf152b4b6d326fe4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 137.0 to 141.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR