5-Bromo-2-iodopyridine ≥97.0%

Leverantör: TCI EUROPE
Varning

B2877-25G B2877-5G
TCIAB2877-25GEA 2275 SEK
TCIAB2877-25G TCIAB2877-5G
5-Bromo-2-iodopyridine ≥97.0%
5-Bromo-2-iodopyridine
Formel: C₅H₃BrIN
MW: 283,89 g/mol
MDL Number: MFCD00691506
CAS nummer: 223463-13-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17c0a1ea80cb90de5912069de5580766a63d1123&vid=17c0a1ea80cb90de5912069de5580766a63d1123&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 113.0 to 117.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR