2-Methyl-1-nitronaphthalene ≥98.0%
Leverantör: TCI EUROPE
Varning
M0376-25G M0376-250G
TCIAM0376-25GEA 885 SEK
TCIAM0376-25G TCIAM0376-250G
2-Methyl-1-nitronaphthalene ≥98.0%
2-Methyl-1-nitronaphthalene
Formel: C₁₁H₉NO₂
MW: 187,2 g/mol
Kokpunkt: 188°C
Smältpunkt: 81 °C
MDL Number: MFCD00003914
CAS nummer: 881-03-8
EINECS: 212-917-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=256ffe12fb9f68ab6ea1570b82988820c29f975c&vid=256ffe12fb9f68ab6ea1570b82988820c29f975c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 83.0 deg-C
Solubility in Methanol almost transparency