1-Naftylamin ≥99.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: 1-Aminonaphthalene
N0052-25G N0052-100G
TCIAN0052-25GEA 685 SEK
TCIAN0052-25G TCIAN0052-100G
1-Naftylamin ≥99.0% (by GC, titration analysis)
1-Naftylamin
Formel: C₁₀H₉N
MW: 143,19 g/mol
Kokpunkt: 301°C
Smältpunkt: 50 °C
Densitet: 1,171 g/cm³ (20 °C)
Flampunkt: 156 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004016
CAS nummer: 134-32-7
EINECS: 205-138-7
UN: 2077
ADR: 6.1,III
Merck Index: 14,06398
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a3c46a74af9744d82694de5de5f4333bef5afdc&vid=1a3c46a74af9744d82694de5de5f4333bef5afdc&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 49.0 to 51.0 deg-C
Solubility in dilute Acetic acid almost transparency