2-Chloronaphthalene ≥98.0%

Leverantör: TCI EUROPE

C0213-1G C0213-25G
TCIAC0213-1GEA 350 SEK
TCIAC0213-1G TCIAC0213-25G
2-Chloronaphthalene ≥98.0%
2-Chloronaphthalene
Formel: C₁₀H₇Cl
MW: 162,62 g/mol
Kokpunkt: 256 °C (1013 hPa)
Smältpunkt: 59,5 °C
Densitet: 1,2656 g/cm³ (16 °C)
MDL Number: MFCD00035731
CAS nummer: 91-58-7
EINECS: 202-079-9
Merck Index: 14,02150
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e48fec82e0bfadd6dd6d7ea1e2fad23e1a2aa8f4&vid=e48fec82e0bfadd6dd6d7ea1e2fad23e1a2aa8f4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR