1-Klornaftalen ≥97.0%

Leverantör: TCI
Varning

Synonyms: Triisodecyl phosphite, α-Chloronaphthalene

C2310-5G C2310-25G
TCIAC2310-5GEA 670 SEK
TCIAC2310-5G TCIAC2310-25G
1-Klornaftalen ≥97.0%
1-Klornaftalen
Formel: C₁₀H₇Cl
MW: 162.62 g/mol
Kokpunkt: 263°C
Smältpunkt: –20°C
Flampunkt: 121°C
Storage Temperature: Omgivande
MDL Number: MFCD00003874
CAS nummer: 90-13-1
EINECS: 201-967-3
UN: 3082
ADR: 9 III
Merck Index: 14,02149
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e586bf747a50aa0f4d10ca02aabe2c077cab493&vid=2e586bf747a50aa0f4d10ca02aabe2c077cab493&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR