2-Naphthaldehyde ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
β-Naphthaldehyde
N0003-25G
N0003-5G
TCIAN0003-25GEA
1150
SEK
TCIAN0003-25G
TCIAN0003-5G
2-Naphthaldehyde ≥98.0%
2-Naphthaldehyde
Formel:
C₁₁H₈O MW: 156,18 g/mol Kokpunkt: 147°C Smältpunkt: 57…60 °C Densitet: 1,0775 g/cm³ (99 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00004094 CAS nummer: 66-99-9 EINECS: 200-640-2 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 60.0 to 64.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...