1-Bromo-4-fluoronaphthalene ≥97.0%
Leverantör: TCI EUROPE
Varning
B3797-1G B3797-5G
TCIAB3797-1GEA 485 SEK
TCIAB3797-1G TCIAB3797-5G
1-Bromo-4-fluoronaphthalene ≥97.0%
1-Bromo-4-fluoronaphthalene
Formel: C₁₀H₆BrF
MW: 225,06 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00051473
CAS nummer: 341-41-3
EINECS: 206-434-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8b884b8c9d1d65cea00a82733a50e50694d28b4&vid=d8b884b8c9d1d65cea00a82733a50e50694d28b4&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Yellow to Orange powder to lump
Purity (GC) min. 97.0 %
Melting point 35.0 to 39.0 deg-C