1-Isoquinolinecarboxylic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: Isoquinoline-1-carboxylic acid

I0671-25G I0671-5G
TCIAI0671-25GEA 1750 SEK
TCIAI0671-25G TCIAI0671-5G
1-Isoquinolinecarboxylic acid ≥98.0% (by GC, titration analysis)
1-Isoquinolinecarboxylic acid
Formel: C₁₀H₇NO₂
MW: 173,17 g/mol
Kokpunkt: 345 °C (1013 hPa)
Smältpunkt: 161 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006901
CAS nummer: 486-73-7
EINECS: 207-639-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e4a0af4765113c0c8fbbb9ea81cc9f4ab49697b&vid=9e4a0af4765113c0c8fbbb9ea81cc9f4ab49697b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR