4-Bromoisoquinoline ≥98.0%

Leverantör: TCI
Varning

B0930-10G B0930-25G
TCIAB0930-10GEA 815 SEK
TCIAB0930-10G TCIAB0930-25G
4-Bromoisoquinoline ≥98.0%
4-Bromoisoquinoline
Formel: C₉H₆BrN
Kokpunkt: 285°C
Storage Temperature: Kylskåp
MDL Number: MFCD00006904
CAS nummer: 1532-97-4
EINECS: 216-244-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5165da99521b20baab44a0568ab08d02c756c7db&vid=5165da99521b20baab44a0568ab08d02c756c7db&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 44.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR