5-Indanol ≥99.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 5-Hydroxyindan, 5-Hydroxyhydrindene, 5-Indanol
H0251-100G H0251-25G
TCIAH0251-100GEA 3275 SEK
TCIAH0251-100G TCIAH0251-25G
5-Indanol ≥99.0%
5-Indanol
Formel: C₉H₁₀O
MW: 134,18 g/mol
Kokpunkt: 255°C
Smältpunkt: 51…53 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003802
CAS nummer: 1470-94-6
EINECS: 216-006-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18e5e0d5477e7b5061d58a457a0cfc10c455ee5f&vid=18e5e0d5477e7b5061d58a457a0cfc10c455ee5f&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 52.0 to 56.0 deg-C
Solubility in Methanol almost transparency