5,6-Dimethoxy-1-indanone ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 5,6-Dimethoxy-2,3-dihydro-1H-inden-1-one
D3313-25G D3313-5G
TCIAD3313-25GEA 895 SEK
TCIAD3313-25G TCIAD3313-5G
5,6-Dimethoxy-1-indanone ≥98.0%
5,6-Dimethoxy-1-indanone
Formel: C₁₁H₁₂O₃
MW: 192,21 g/mol
Kokpunkt: 139°C
Smältpunkt: 120 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003790
CAS nummer: 2107-69-9
EINECS: 218-287-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d441f08e255428ddc450741be00b5c1acf74a7d&vid=2d441f08e255428ddc450741be00b5c1acf74a7d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 118.0 to 122.0 deg-C