5-Methoxy-1-indanone ≥98.0%
Leverantör: TCI
M1467-25G
M1467-5G
TCIAM1467-25GEA
1950
SEK
TCIAM1467-25G
TCIAM1467-5G
5-Methoxy-1-indanone ≥98.0%
5-Methoxy-1-indanone
Formel:
C₁₀H₁₀O₂ Kokpunkt: 150°C Smältpunkt: 107…108 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00003789 CAS nummer: 5111-70-6 EINECS: 225-838-6 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 108.0 to 112.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...