2-Indanone ≥98.0%

Leverantör: TCI

Synonyms: beta-Hydrindone, 2-Oxoindane, 1H-Inden-2(3H)-one

I0649-25G I0649-5G
TCIAI0649-25GEA 1275 SEK
TCIAI0649-25G TCIAI0649-5G
2-Indanone ≥98.0%
2-Indanone
Formel: C₉H₈O
Kokpunkt: 225°C
Smältpunkt: 54…56 °C
Densitet: 1,0712 g/cm³ (67 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00003792
CAS nummer: 615-13-4
EINECS: 210-410-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=215cffa254a48fc822b9fd240434bbda83ecde41&vid=215cffa254a48fc822b9fd240434bbda83ecde41&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR