Diphenyl ditelluride ≥98.0%

Leverantör: TCI
Fara

D2718-1G D2718-5G
TCIAD2718-1GEA 925 SEK
TCIAD2718-1G TCIAD2718-5G
Diphenyl ditelluride ≥98.0%
Diphenyl ditelluride
Formel: C₁₂H₁₀Te₂
MW: 409.41 g/mol
Smältpunkt: 67
MDL Number: MFCD00192106
CAS nummer: 32294-60-3
EINECS: 250-982-1
UN: 3284
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2e22e5ff1f280fc3d8142d032c662237b611e38&vid=a2e22e5ff1f280fc3d8142d032c662237b611e38&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 67.0 deg-C
Solubility in Toluene within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR