Diphenyl diselenide ≥97.0%
Leverantör: TCI
|
Fara
|
D2186-25G
D2186-5G
TCIAD2186-25GEA
1550
SEK
TCIAD2186-25G
TCIAD2186-5G
Diphenyl diselenide ≥97.0%
Diphenyl diselenide
Formel:
C₁₂H₁₀Se₂ MW: 312.13 g/mol Kokpunkt: 202°C Storage Temperature: Omgivande |
MDL Number:
MFCD00003001 CAS nummer: 1666-13-3 EINECS: 216-780-2 UN: 3283 ADR: 6.1 II |
|
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 60.0 to 63.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...