Dimethyl diselenide ≥98.0%

Leverantör: TCI
Fara

Synonyms: Dimethyldiselenide

D3290-1G
TCIAD3290-1GEA 1050 SEK
TCIAD3290-1G
Dimethyl diselenide ≥98.0%
Dimethyl diselenide
Formel: C₂H₆Se₂
MW: 187.99 g/mol
Kokpunkt: 155°C
Smältpunkt: –82°C
Densitet: 2.0009
Flampunkt: 60°C
MDL Number: MFCD00008254
CAS nummer: 7101-31-7
EINECS: 230-410-7
UN: 3440
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=720a13a923117e47258e6dc649f8ddc620c7fd40&vid=720a13a923117e47258e6dc649f8ddc620c7fd40&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR