Tetraklorsilan ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Kiseltetraklorid

T0636-100G T0636-25G T0636-500G
TCIAT0636-100GEA 365 SEK
TCIAT0636-100G TCIAT0636-25G TCIAT0636-500G
Tetraklorsilan ≥98.0%
Tetraklorsilan
Formel: Cl₄Si
MW: 169,9 g/mol
Kokpunkt: 59°C
Smältpunkt: –68°C
Densitet: 1,48 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011229
CAS nummer: 10026-04-7
EINECS: 233-054-0
UN: 1818
ADR: 8,II
Merck Index: 14,08500
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=994aadf6eebc7ffabda85cae1f8ba7a1a7b7f0e9&vid=994aadf6eebc7ffabda85cae1f8ba7a1a7b7f0e9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR