Methyl-6-methylnicotinate ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Methyl 6-methylnicotinate
M1893-25G
M1893-5G
TCIAM1893-25GEA
2475
SEK
TCIAM1893-25G
TCIAM1893-5G
Methyl-6-methylnicotinate ≥98.0% (by GC, titration analysis)
Methyl-6-methylnicotinate
Formel:
C₈H₉NO₂ MW: 151,16 g/mol Kokpunkt: 160°C |
MDL Number:
MFCD00006340 CAS nummer: 5470-70-2 |
Specifikation testresultat
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...