Ethyl-6-chloronicotinate ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Ethyl 6-chloronicotinate

E0852-25G E0852-5G
TCIAE0852-25GEA 935 SEK
TCIAE0852-25G TCIAE0852-5G
Ethyl-6-chloronicotinate ≥97.0%
Ethyl-6-chloronicotinate
Formel: C₈H₈ClNO₂
MW: 185,61 g/mol
Kokpunkt: 150°C
Storage Temperature: Kylskåp
MDL Number: MFCD00082739
CAS nummer: 49608-01-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=918421d169e65562de5f52f4e38ffb287f86fb97&vid=918421d169e65562de5f52f4e38ffb287f86fb97&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR