Diethyl chlorophosphate ≥97.0%
Leverantör: TCI EUROPE
Fara
D2206-25G D2206-250G
TCIAD2206-25GEA 390 SEK
TCIAD2206-25G TCIAD2206-250G
Diethyl chlorophosphate ≥97.0%
Diethyl chlorophosphate
Formel: C₄H₁₀ClO₃P
MW: 172,55 g/mol
Kokpunkt: 93°C
Flampunkt: 61°C
MDL Number: MFCD00009075
CAS nummer: 814-49-3
EINECS: 212-396-4
UN: 2927
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3a71b91f3fa5f3bc162607bd03abf2d5fa46b4e&vid=c3a71b91f3fa5f3bc162607bd03abf2d5fa46b4e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Argentmetric Titration) min. 97.0 %