Ethylene chlorophosphite ≥97.0%
Leverantör: TCI EUROPE
Fara
C1215-25G C1215-5G
TCIAC1215-25GEA 845 SEK
TCIAC1215-25G TCIAC1215-5G
Ethylene chlorophosphite ≥97.0%
Ethylene chlorophosphite
Formel: C₂H₄ClO₂P
MW: 126,48 g/mol
Kokpunkt: 47°C
Smältpunkt: -28°C
Flampunkt: 70°C
Storage Temperature: Kylskåp
MDL Number: MFCD00014511
CAS nummer: 822-39-9
EINECS: 212-499-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cd632fc8246ab87b704cc36cbba690a221082e5&vid=7cd632fc8246ab87b704cc36cbba690a221082e5&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %