2-Amino-2-thiazoline hydrochloride ≥98.0%

Leverantör: TCI EUROPE

A0936-25G A0936-5G
TCIAA0936-25GEA 1950 SEK
TCIAA0936-25G TCIAA0936-5G
2-Amino-2-thiazoline hydrochloride ≥98.0%
2-Amino-2-thiazoline hydrochloride
Formel: C₃H₆N₂S
MW: 102,16 g/mol
MDL Number: MFCD00012709
CAS nummer: 3882-98-2
EINECS: 223-416-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3e72137bf0573e148fe09a2122fc950df8f61cc&vid=f3e72137bf0573e148fe09a2122fc950df8f61cc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 198.0 to 204.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR