Bis(cyclopentadienyl)zirconium dichloride ≥97.0%
Leverantör: TCI
|
Fara
|
Synonyms:
Zirconocene dichloride
Z0007-25G
Z0007-5G
TCIAZ0007-25GEA
1750
SEK
TCIAZ0007-25G
TCIAZ0007-5G
Bis(cyclopentadienyl)zirconium dichloride ≥97.0%
Bis(cyclopentadienyl)zirconium dichloride
Formel:
C₁₀H₁₀Cl₂Zr MW: 292.32 g/mol |
MDL Number:
MFCD00003726 CAS nummer: 1291-32-3 EINECS: 215-066-8 UN: 3261 ADR: 8 III |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...