Methylenediphosphonic acid ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Medronic acid

M0843-1G M0843-5G
TCIAM0843-1GEA 615 SEK
TCIAM0843-1G TCIAM0843-5G
Methylenediphosphonic acid ≥97.0%
Methylenediphosphonic acid
Formel: CH₆O₆P₂
MW: 176 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00002139
CAS nummer: 1984-15-2
EINECS: 217-851-0
UN: 3261
ADR: 8,III
Merck Index: 14,05792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf670a87237580e2143ead571124340c8504446f&vid=bf670a87237580e2143ead571124340c8504446f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 97.0 %
Melting point 201.0 to 207.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR