Octaethylene glycol ≥96.0%

Leverantör: TCI EUROPE

Synonyms: PEG-08, 3,6,9,12,15,18,21-Heptaoxatricosane-1,23-diol

O0295-1G O0295-5G
TCIAO0295-1GEA 885 SEK
TCIAO0295-1G TCIAO0295-5G
Octaethylene glycol ≥96.0%
Octaethylene glycol
Formel: C₁₆H₃₄O₉
MW: 370,44 g/mol
MDL Number: MFCD00698694
CAS nummer: 5117-19-1
EINECS: 225-856-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c1b84d0b0610ea109cf28075e4304c281a4a645&vid=3c1b84d0b0610ea109cf28075e4304c281a4a645&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR