N,N-Dimethylglycine methyl ester ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
D0257-25G D0257-500G D0257-5G
TCIAD0257-25GEA 910 SEK
TCIAD0257-25G TCIAD0257-500G TCIAD0257-5G
N,N-Dimethylglycine methyl ester ≥98.0% (by GC, titration analysis)
N,N-Dimethylglycine methyl ester
Formel: C₅H₁₁NO₂
Kokpunkt: 135°C
Storage Temperature: Frys
MDL Number: MFCD00059303
CAS nummer: 7148-06-3
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0288f5dfc4f3aa513ec9dbeeeacc1c6c891f6a9d&vid=0288f5dfc4f3aa513ec9dbeeeacc1c6c891f6a9d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %