Diethyl acetamidomalonate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Diethyl 2-acetamidomalonate

A0015-25G A0015-500G A0015-100G
TCIAA0015-25GEA 215 SEK
TCIAA0015-25G TCIAA0015-500G TCIAA0015-100G
Diethyl acetamidomalonate ≥98.0%
Diethyl acetamidomalonate
Formel: C₉H₁₅NO₅
MW: 217,22 g/mol
MDL Number: MFCD00009146
CAS nummer: 1068-90-2
EINECS: 213-952-9

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 95.0 to 99.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR